Order Entry
Belgium
ContactUsLinkComponent
Sodium carbonate, Test Solution (TS)
Irritant
Sodium carbonate, Test Solution (TS)
Catalog # ACSDCH8002113.L1
Supplier:  CPACHEM
New Product
CAS Number:  
Irritant
Sodium carbonate, Test Solution (TS)
Catalog # ACSDCH8002113.L1
Supplier:  CPACHEM
New Product
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :105.99 g/mol
EINECS:207-838-8
 :GHS07
Storage Temperature:Room Temperature
CAS Number:497-19-8
Formula:Na₂CO₃
MDL Number:MFCD00003494