Order Entry
Belgium
ContactUsLinkComponent
Potassium carbonate, Test Solution (TS)
Potassium carbonate, Test Solution (TS)
Catalog # ACSDCH8002100.L1
Supplier:  CPACHEM
New Product
CAS Number:  
Potassium carbonate, Test Solution (TS)
Catalog # ACSDCH8002100.L1
Supplier:  CPACHEM
New Product
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :138.21 g/mol
EINECS:209-529-3
CAS Number:584-08-7
Formula:K₂CO₃
MDL Number:MFCD00011382