Order Entry
Belgium
ContactUsLinkComponent
Nickel, ICP-MS Standard Solution
Nickel, ICP-MS Standard Solution
Catalog # ACSDM238.2NP.L1
Supplier:  CPACHEM
New Product
CAS Number:  
Nickel, ICP-MS Standard Solution
Catalog # ACSDM238.2NP.L1
Supplier:  CPACHEM
New Product
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HNO₃
  • Pk:
    100 mL
  • Environmentally Preferable:
  • Element:
    Nickel
  • Symbol:
    Ni

Specifications

About this item

Certifications: ISO 17034, ISO 17025, ISO 9001