Order Entry
Belgium
Orders LinkContactUsLinkComponent
Potassium Dichromate Solution for Absorbance Control

Potassium Dichromate Solution for Absorbance Control

2×10 ml Potassium dichromate solution for absorbance control 235 - 350 nm and 6×10 ml, 80 mL
Catalog # ACSDUV2225PDA.L08
Supplier:   CPACHEM
Potassium Dichromate Solution for Absorbance Control
Catalog # ACSDUV2225PDA.L08
Supplier:  CPACHEM
CAS Number:  
Potassium Dichromate Solution for Absorbance Control

2×10 ml Potassium dichromate solution for absorbance control 235 - 350 nm and 6×10 ml, 80 mL

CAS Number:  
Price
202,00 €
Log in for availability
1 * 80 mL
Product Details & Documents

Certifications: ISO 17034, 17025, 9001 certified.

Specifications

Specifications

  • Pk:
    80 mL
  • Description:
    2×10 ml Potassium dichromate solution for absorbance control 235 - 350 nm and 6×10 ml
  • Cat. No.:
    ACSDUV2225PDA.L08
  • Storage conditions:
    36 months
  • Storage temperature:
    15...25 °C

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.