Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Cationic Standards
Iron Standards
Iron Standard Stock Solution
Iron Standard Stock Solution
ACSDCH0807.L1
:
CPACHEM
New Product
:
Iron Standard Stock Solution
ACSDCH0807.L1
:
CPACHEM
New Product
:
Pk:
100 mL
Description:
Iron standard stock solution
Storage temperature:
15...25 °C
:
ISO 9001 certified.