Check Valve
Check valve with loose closure and variable weight, without spindle valve, 14,5/23 NS core, Glass
Catalog # NEUB0730-20-14
Supplier: NEUBERT VOLUME GLASSWAERE
Jump to:
Product Details & Documents
Loose closure with variable weight check valve.
- Ball grinding closure with sealing
- Dead weight of the ground hood
- Adjustable by additional glass weights
Material: Borosilicate glass 3.3; filled with glass beads; with closure cap GL 32.
Specifications
- Housing material:Glass
- Description:Check valve with loose closure and variable weight, without spindle valve, 14,5/23 NS core