ContactUsLinkComponent
Beaded Edge, PTFE Faucet
Catalog # NEUB1195-55-25250
New Product
Beaded Edge, PTFE Faucet
Catalog # NEUB1195-55-25250
New Product
Specifications
  • Description:
    Beaded edge, PTFE faucet
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
Beaded edge PTFE faucet with punctures.
  • Borosilicate 3.3
  • PTFE tap NS 14,5/2,5 mm
Internal diameter: Ø 25 mm
Length: 250 mm
Tube: Ø 28×1,4 mm