ContactUsLinkComponent
AlphaTec® 04-005 Long-Cuffed PVC Gloves
AlphaTec® 04-005 Long-Cuffed PVC Gloves
Supplier:  Ansell
New Product
Certificates
undefined
AlphaTec® 04-005 Long-Cuffed PVC Gloves
Supplier:  Ansell
New Product
Specifications
  • Catalog No:
  • ANSE04005/8
  • ANSE04005/9
  • ANSE04005/10
  • ANSE04005/11
  • Latex / latex-free:
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Material:
  • PVC/nitrile
  • PVC/nitrile
  • PVC/nitrile
  • PVC/nitrile
  • Finish:
  • Foam
  • Foam
  • Foam
  • Foam
  • Cuff style:
  • Gauntlet
  • Gauntlet
  • Gauntlet
  • Gauntlet
  • Palm thickness:
  • 1,80 mm
  • 1,80 mm
  • 1,80 mm
  • 1,80 mm
  • Anti-static:
  • +
  • +
  • +
  • +
  • Food contact approved:
  • +
  • +
  • +
  • +