ContactUsLinkComponent
Preventive Maintenance (PM) Kit
Preventive Maintenance (PM) Kit
Catalog # VELP40003081
Supplier:  VELP SCIENTIFIC
New Product
Preventive Maintenance (PM) Kit
Catalog # VELP40003081
Supplier:  VELP SCIENTIFIC
New Product
Specifications
  • Description:
    PM kit includes 4× ferule, 1× top reactor gasket Ø 18 mm, 3× gasket for autosampler piston, 2,5× Teflon tube AWG16 TW (VELP10004795), 8× ferule for VELP10004795 (VELP10004929), 3× OR 4030 viton, 4× ss tubing Ø 2×1 mm, 1× fan 80×80×25, 24 V
  • Model:
    EMA502
  • Pk:
    1 KIT
Specifications
About this item
The PM full is designed to keep your running with high performances while ensuring long life and reliability, even under high workloads.
  • Reduction of unplanned downtime with consequent improvement of profit and production performance
  • Lower costs for the purchase of necessary spare parts and eventual extraordinary maintenance
  • Reduction of lost time associated with unplanned service intervals and instrument downtime
  • All the necessary spare parts from the manufacturer are always at hand