Order Entry
Belgium
ContactUsLinkComponent
Middlebrook 7H10 OADC (Oleic Albumin Dextrose Catalase) Growth Supplement
Middlebrook 7H10 OADC (Oleic Albumin Dextrose Catalase) Growth Supplement
Catalog # USBIM3895-01-500G
Supplier:  US Biological
New Product
CAS Number:  
Middlebrook 7H10 OADC (Oleic Albumin Dextrose Catalase) Growth Supplement
Catalog # USBIM3895-01-500G
Supplier:  US Biological
New Product
CAS Number:  

Specifications

  • Pk:
    500 g
  • Colour:
    Beige/green
  • Description:
    Middlebrook 7H10 OADC (Oleic albumin dextrose catalase) growth supplement
  • Storage temperature:
    4 °C
  • Contents:
    Bovine albumin fraction V 5,0 g/100 ml
    Catalase 0,004 g/100 ml
    D-glucose 2,0 g/100 ml
    Oleic acid 0,05 g/100 ml
    Sodium chloride 0,85 g/100 ml
  • Formulation:
    OADC Enrichment medium is used with Middlebrook 7H10 Mycobacteria agar (Powder)
  • Grade:
    Microbiological grade
  • Appearance:
    Light beige with slight green tint, homogeneous, free flowing powder
  • Form:
    Powder

Specifications

About this item

Enrichment supplement recommended for the cultivation of Mycobacteria sp. OADC enrichment medium is used with Middlebrook 7H10 Mycobacteria Agar (Powder).

  • Light beige with slight green tint
  • Homogeneous
  • Free flowing powder

Solubility: Light yellow, turbid.