Nessler reagent solutions (I+II) [Potassium Iodomercurate solution + NaOH 25% w/v solution] Reag. Ph. Eur. 1071600 for determination of ammonia and ammonium salts
Catalog # 87709.200
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:200 mL
- Seq:2530
- Pk Info:100 ml + 100 ml
Specifications
Specification Test Results
- Performance test:Passes test
Specification Test Results
About this item
:
:786.4
ADR:8,II
CAS Number:7783-33-7