ContactUsLinkComponent
4-Nitrophenyl phosphate disodium salt hexahydrate (pNPP)
  ROTH4165.1
 :  Roth Carl
 :  
4-Nitrophenyl phosphate disodium salt hexahydrate (pNPP)
  ROTH4165.1
 :  Roth Carl
 :  
  • Pack type:
    Glass bottle for solids
  • Pk:
    2,5 g
 :
 :300 °C
 :371.144 g/mol
 :Freezer
 :224-246-5
 :333338-18-4
 :4264-83-9
 :C₆H₄NNa₂O₆P·6H₂O
 :MFCD00066288