N-Octyl-β-D-glucopyranoside non-ionic detergent, Pierce™
Catalog # PIER28310
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
- Environmentally Preferable:
Specifications
About this item
Synonyms: Octylglucopyranoside, OG, (2R,3S,4S,5R,6R)-2-(Hydroxymethyl)-6-(octyloxy)tetrahydro-2H-pyran-3,4,5-triol, n-Octyl β-D-glucopyranoside, 1-O-n-Octyl-β-D-glucopyranoside, Octyl-β-D-glucopyranoside
:
:292.37 g/mol
Melting Pt:110 °C
EINECS:249-887-8
:GHS07
Storage Temperature:Freezer
CAS Number:29836-26-8
Formula:C₁₄H₂₈O₆
MDL Number:MFCD00063288