


Formaldehyde 37% in aqueous solution
Catalog # FFCHFF1130250
Supplier: FF CHEMICALS
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:250 mL
- Seq:2590
Specifications
About this item
:
:30.03 g/mol
Storage Temperature:Ambient
EINECS:200-001-8
UN:2209
ADR:8,III
:GHS06
:GHS05
:GHS08
CAS Number:50-00-0
Formula:CH₂O
MDL Number:MFCD00003274