
Silica gel, Pore size: 80 Å ≥99.99%, spherical 2.2 µm APS for HPLC
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:63231-67-4
:7631-86-9
Formula:SiO₂
MDL Number:MFCD00011232
:60.08 g/mol
EINECS:231-545-4
:GHS08
Storage Temperature:Ambient
Related Information
Frequently Bought Together