Intelsius® PharmaTherm Temperature Controlled Packaging
About this item
Utilises pharmachill cool packs are hard wearing and durable re-usable coolant, reducing costs and landfill waste.
2 to 8 °C systems qualified against the Intelsius high performance quad profile with stress peaks and overlapping mid season temperatures, the PharmaTherm offers robust performance for all season shipping and peace of mind.
15 to 25 °C systems qualified against Intelsius controlled room temperatures profiles for ambient shipments. GDP compliant systems.
4 Options Available Below
Product Details & Documents
Utilises pharmachill cool packs are hard wearing and durable re-usable coolant, reducing costs and landfill waste.
2 to 8 °C systems qualified against the Intelsius high performance quad profile with stress peaks and overlapping mid season temperatures, the PharmaTherm offers robust performance for all season shipping and peace of mind.
15 to 25 °C systems qualified against Intelsius controlled room temperatures profiles for ambient shipments. GDP compliant systems.
Product Family Options
High perfomance quad profiles
Product Information
- CapacityW×D×H ext.W×D×H int.AvailabilityPrice
- DGINPHT001Pharmatherm high performance, holding time: 96 hr, weight: 5,5 kgPharmatherm high performance, holding time: 96 hr, weight: 5,5 kg5,2 L396×396×396 mm145×225×160 mmLog in for availability1 * 1 ST58,20 €
Controlled room temperatures profiles
Product Information
- CapacityW×D×H ext.W×D×H int.AvailabilityPrice
- DGINPHT019Pharmatherm controlled room temperature, holding time: 96 hr, weight: 7,6 kgPharmatherm controlled room temperature, holding time: 96 hr, weight: 7,6 kg6,6 L396×396×396 mm189×189×185 mmLog in for availability1 * 1 ST66,90 €
- DGINPHT020Pharmatherm controlled room temperature, holding time: 96 hr, weight: 11,4 kgPharmatherm controlled room temperature, holding time: 96 hr, weight: 11,4 kg15,2 L460×460×460 mm249×249×245 mmLog in for availability1 * 1 ST90,20 €
- DGINPHT021Pharmatherm controlled room temperature, holding time: 96 hr, weight: 18,1 kgPharmatherm controlled room temperature, holding time: 96 hr, weight: 18,1 kg26,8 L565×565×565 mm374×319×225 mmLog in for availability1 * 1 ST135,00 €