Human Recombinant ASD3 (MYH6) (NM_002471)
Catalog # ORIGTP313673
Supplier: OriGene
New Product
CAS Number:
Specifications
- Pk:20 µG
- Protein/peptide type:Recombinant
- Source:HEK293T cells
- Species:Human
- Tag sequence:C-Myc/DDK
- Storage conditions:25 mM Tris-HCl, 100 mM glycine, pH 7,3, 10% glycerol. Store at −80 °C.
- Protein synonyms:CMH14|MYHCA|MYHC|ASD3|alpha-MHC|CMD1EE|SSS3
- UniProtKB:P13533
- Protein/peptide name:ASD3 (MYH6) (NM_002471)
- Purity:>80% as determined by SDS-PAGE and Coomassie blue staining
- Molecular weight:223,6 kDa
- Concentration:>0,05 µg/µl as determined by microplate BCA method
Specifications
About this item
Purified recombinant protein of Homo sapiens myosin, heavy chain 6, cardiac muscle, alpha (MYH6).
For testing in cell culture applications, please filter before use. Some loss of protein during the filtration process may occur.
Stable for 12 months from the date of receipt of the product under proper storage and handling conditions. Avoid repeated freeze-thaw cycles.