Order Entry
Belgium
ContactUsLinkComponent
Tetrazolium blue chloride for colorimetric determination of reducing compounds, Sigma-Aldrich®
Tetrazolium blue chloride for colorimetric determination of reducing compounds, Sigma-Aldrich®
Certificates
undefined
Tetrazolium blue chloride for colorimetric determination of reducing compounds, Sigma-Aldrich®

Specification Test Results

  • Appearance (Colour):
    Yellow
  • Appearance (Form):
    Powder
  • Solubility (Colour):
    Yellow to Yellow-Green
  • Solubility (Turbidity)
    50 mg/mL, MeOH:
    Clear
  • Infrared spectrum:
    Conforms to Structure
  • Carbon:
    63.4 - 67.4 %
  • Nitrogen:
    14.9 - 15.9 %

Specification Test Results

Frequently Bought Together