
Nitro blue tetrazolium chloride (NBT)
Catalog # SOUT0204-01
Supplier: SOUTHERN BIOTECHNOLOGY
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:200 mg
- Seq:2530
Specifications
About this item
:
:817.65 g/mol
Melting Pt:200 °C
EINECS:206-067-4
:GHS07
Storage Temperature:Refrigerator
CAS Number:298-83-9
Formula:C₄₀H₃₀Cl₂N₁₀O₆
MDL Number:MFCD00012159