Order Entry
Belgium
ContactUsLinkComponent
 
 
 
Nitro blue tetrazolium chloride (NBT) ≥90% (by HPLC), Sigma-Aldrich®
 
undefined
 
 
Nitro blue tetrazolium chloride (NBT) ≥90% (by HPLC), Sigma-Aldrich®
 

 

  • Appearance (Colour):
    Yellow
  • Appearance (Form):
    Powder
  • Infrared spectrum:
    Conforms to Structure
  • Water (by Karl Fischer):
    ≤4.5 %
  • Solvent Content (by GC):
    ≤10.0 %
  • Nitrogen
    Corrected for Water and Solvent:
    16.4 - 17.9 %
  • Purity (HPLC):
    ≥90.0 %
  • Solubility (Colour)
    c = 10 mg/ml; Water:
    Green-Yellow to Dark Yellow
  • Solubility (Turbidity):
    Clear
  • Recommended Retest Period:
    3 Years

 

 

Frequently Bought Together