

Nitro blue tetrazolium chloride (NBT), tablets, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:298-83-9
:
Formula:C₄₀H₃₀Cl₂N₁₀O₆
MDL Number:MFCD00012159
:817.65 g/mol
Melting Pt:200 °C
:M-Clarity™ quality level = MQ200
EINECS:206-067-4
:GHS05
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance (Colour):Yellow
- Diameter:5/32 inches
- Form:Round Tablet
- Dissolution Time:≤5 min
- Solubility (Colour):Faint Yellow to Yellow
- Solubility (Turbidity):Clear
- pH:4.5 - 5.5
- Total Tablet Weight:21.2 - 28.8 mg/tab
- Note:pH Test,Dissolution time, and solubility are the results from one tablet in one ml of water.
Specification Test Results
Frequently Bought Together