(3-((3-Cholamidopropyl)dimethylammonio)-2-hydroxy-1-propanesulphonate), Millipore®
Supplier: Merck Millipore (Calbiochem)
Certificates
About this item
CAS Number:82473-24-3
:
Formula:C₃₂H₅₈N₂O₈S
:630.89 g/mol
Melting Pt:184…186 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together