Order Entry
Belgium
Orders LinkContactUsLinkComponent
Caffeine, Supelco®

Caffeine, Supelco®

Glass bottle for solids, 5 g
Catalog # 75035-5G-F
Irritant
Supplier:   Merck
Caffeine, Supelco®
Catalog # 75035-5G-F
Supplier:  Merck
CAS Number:  58-08-2
Caffeine, Supelco®
Caffeine, Supelco®

Glass bottle for solids, 5 g

CAS Number:  58-08-2
Price
53,90 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  3,7-Dihydro-1,3,7-trimethyl-1H-purine-2,6-dione, 7-Methyltheobromine, 1,3,7-Trimethylxanthine, Methyltheobromine, 1,3,7-Trimethyl-2,6-dioxopurine, 2,6-Dihydroxy-1,3,7-trimethylpurine
  • CAS Number:58-08-2
  • Formula:C₈H₁₀N₄O₂
  • MDL Number:MFCD00005758
  • Molecular Weight:194.19 g/mol
  • Merck Index:13,01636
  • Melting Pt:235…239 °C
  • Density:1.23 g/cm³ (19 °C)
  •  :M-Clarity™ quality level = MQ100
  • Storage Temperature:Ambient
  • EINECS:200-362-1

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.