
N,N-Diethyl-p-phenylenediammonium sulphate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:6283-63-2
:
Formula:C₁₀H₁₈N₂O₄S
MDL Number:MFCD00012993
:262.33 g/mol
Boiling Pt:274 - 275 °C (1013 hPa)
Melting Pt:184…186 °C
Storage Temperature:Ambient
EINECS:228-500-6
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 % (calcd.on dried substance)
- Solubility in Water:almost transparency
- Drying loss:max. 5.0 %
Specification Test Results
Frequently Bought Together