Order Entry
Belgium
ContactUsLinkComponent
 
 
EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride), Pierce™
 
undefined
 
EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride), Pierce™
 

 

  • Product Line:
    Pierce™
  • Labeling Method:
    Chemical Labeling
  • Crosslinker Type:
    Heterobifunctional
  • Reactive Moiety:
    Carbodiimide
  • Spacer:
    Short
  • Form:
    Powder
  • Quantity:
    10 mg/5 g/25 g
  • Type:
    Crosslinker
  • Solubility:
    Water

 

 

Frequently Bought Together