Order Entry
Belgium
ContactUsLinkComponent
NNN Modified Medium, Twin Pack, Dehydrated
NNN Modified Medium, Twin Pack, Dehydrated
Supplier:  HIMEDIA
New Product
Certificates
NNN Modified Medium, Twin Pack, Dehydrated
Supplier:  HIMEDIA
New Product

Specifications

  • Form:
    Powder
  • Storage temperature:
    15…25 °C

Specifications

Frequently Bought Together