Order Entry
Belgium
Orders LinkContactUsLinkComponent
Aspartame

Aspartame

Supplier:   Spectrum Chemical
Aspartame
Supplier:  Spectrum Chemical
Certificates

2 Options Available Below

Product Details & Documents

Aspartame, Powder, NF. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).

  • Transmittance at 430 nm: minimum 0,95
  • Specific rotation (A)D: +14,5 to +16,5
  • Loss On drying: Max. 4,5%
  • Residue on ignition: Max. 0,2%
Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • SPCMA1378-500GM
  • SPCMA1378-2.5KG
  • Protein/peptide name:
  • Aspartame
  • Aspartame
  • Protein synonyms:
  • Methyl aspartylphenylalanate|stereoisomer|L-Phenylalanine, N-L-alpha-aspartyl, 1-methyl ester|N-L-alpha Aspartyl-L-phenlalanine 1-methyl ester|Succinamic acid|Nutrasweet|N-methyl ester|3-amino-N-(alpha-carboxyphenethyl)-|1-Methyl N-L-alpha-aspartyl-L-phenylalanine|Aspartylphenylalanine methyl ester|3-Amino-N-(alpha-carboxyphenethyl) succinamic acid N-methyl ester
  • Methyl aspartylphenylalanate|stereoisomer|L-Phenylalanine, N-L-alpha-aspartyl, 1-methyl ester|N-L-alpha Aspartyl-L-phenlalanine 1-methyl ester|Succinamic acid|Nutrasweet|N-methyl ester|3-amino-N-(alpha-carboxyphenethyl)-|1-Methyl N-L-alpha-aspartyl-L-phenylalanine|Aspartylphenylalanine methyl ester|3-Amino-N-(alpha-carboxyphenethyl) succinamic acid N-methyl ester
  • CAS No.:
  • 22839-47-0
  • 22839-47-0
  • MDL No.:
  • MFCD00002724
  • MFCD00002724
  • Purity:
  • 98,0% (Dried basis)
  • 98,0% (Dried basis)
  • Grade:
  • NF
  • NF
  • Form:
  • Powder
  • Powder
  • Molecular weight:
  • 294,30 g/mol
  • 294,30 g/mol
  • Molecular formula:
  • C₁₄H₁₈N₂O₅
  • C₁₄H₁₈N₂O₅
  • Storage conditions:
  • Ambient
  • Ambient

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Poly pail
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 933,00 €
    933,00 €
    1 * 500 g
       933,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    SPCMA1378-500GM
    Protein/peptide name
    Aspartame
    Protein synonyms
    Methyl aspartylphenylalanate|stereoisomer|L-Phenylalanine, N-L-alpha-aspartyl, 1-methyl ester|N-L-alpha Aspartyl-L-phenlalanine 1-methyl ester|Succinamic acid|Nutrasweet|N-methyl ester|3-amino-N-(alpha-carboxyphenethyl)-|1-Methyl N-L-alpha-aspartyl-L-phenylalanine|Aspartylphenylalanine methyl ester|3-Amino-N-(alpha-carboxyphenethyl) succinamic acid N-methyl ester
    CAS No.
    22839-47-0
    MDL No.
    MFCD00002724
    Purity
    98,0% (Dried basis)
    Grade
    NF
    Form
    Powder
    Molecular weight
    294,30 g/mol
    Molecular formula
    C₁₄H₁₈N₂O₅
    Storage conditions
    Ambient
  • Poly bottle
    2,5 kg
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 1 290,00 €
    1 290,00 €
    1 * 2,5 kg
       1 290,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    SPCMA1378-2.5KG
    Protein/peptide name
    Aspartame
    Protein synonyms
    Methyl aspartylphenylalanate|stereoisomer|L-Phenylalanine, N-L-alpha-aspartyl, 1-methyl ester|N-L-alpha Aspartyl-L-phenlalanine 1-methyl ester|Succinamic acid|Nutrasweet|N-methyl ester|3-amino-N-(alpha-carboxyphenethyl)-|1-Methyl N-L-alpha-aspartyl-L-phenylalanine|Aspartylphenylalanine methyl ester|3-Amino-N-(alpha-carboxyphenethyl) succinamic acid N-methyl ester
    CAS No.
    22839-47-0
    MDL No.
    MFCD00002724
    Purity
    98,0% (Dried basis)
    Grade
    NF
    Form
    Powder
    Molecular weight
    294,30 g/mol
    Molecular formula
    C₁₄H₁₈N₂O₅
    Storage conditions
    Ambient
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.