Orders LinkContactUsLinkComponent
Quantitative filter papers, ashless grades, MN 640
Quantitative filter papers, ashless grades, MN 640
Fournisseur:  MACHEREY-NAGEL
Certificats
Quantitative filter papers, ashless grades, MN 640
63 Options disponibles ci-dessous
Product Details & Documents
These quantitative filter papers are made from refined pulp and linters. They are purified in a highly developed clean-up process to remove organic and inorganic impurities. This ensures a low ash content of <0,01 %. They are consistent performance and give safe results in gravimetric analyses.
  • Very low ash content
  • Suitable for gravimetric, quantitative analysis
  • Contamination-free filtration
Grade MN 640 dd 'green ribbon' is an ashless, slow filter paper which is slightly less retentive than MN 640 d.
Grade MN 640 d 'blue ribbon' is an ashless, slow filter paper with very fine particle retention. Standard paper for critical gravimetric analysis of fine precipitates like barium sulfate or metastannic acid.
Grade MN 640 de is an ashless, very slow filter paper for the filtration of extremely fine particles in quantitative range.
Grade MN 640 m 'white ribbon' is the classic, general purpose, ashless filter paper with medium speed and medium retention. Typical applications include gravimetric analysis for components, quantitative determinations of sediments and high purity clean up procedures.
Grade MN 640 md 'red ribbon' is an ashless filter paper, which is slightly more retentive than MN 640 m with corresponding increase in filtration time.
Grade MN 640 we is the fastest quantitative filter paper with highest flow rate. It is used for example in analytical procedures of samples with coarse or gelatinous particles.
Grade MN 640 w 'black ribbon' is a fast, ashless filter paper. Recommended for analytical procedures involving coarse particles or gelatinous precipitates.
Spécifications de la famille de produits
Spécifications
  • Catalog No:
  • MANA205005
  • 512-0495
  • 512-0496
  • 512-0497
  • 515-9800
  • MANA205015
  • MANA205018
  • MANA205024
  • MANA205032
  • MANA206005
  • MANA206007
  • MANA206009
  • MANA206011
  • MANA206012
  • MANA206015
  • MANA206018
  • MANA206024
  • MANA206032
  • MANA207005
  • MANA207007
  • MANA207009
  • MANA207011
  • MANA207012
  • MANA207015
  • MANA207018
  • MANA207024
  • MANA207032
  • MANA203005
  • 512-0490
  • MANA203009
  • 512-0491
  • 515-9710
  • 512-0492
  • 512-0493
  • MANA203024
  • MANA203032
  • MANA204005
  • MANA204007
  • MANA204009
  • MANA204011
  • 512-0494
  • MANA204015
  • MANA204018
  • MANA204024
  • MANA204032
  • 512-0084
  • 512-0486
  • 515-9700
  • 512-0487
  • 515-9701
  • 512-0488
  • MANA202018
  • 512-0489
  • MANA202032
  • MANA201005
  • MANA201007
  • MANA201009
  • MANA201011
  • MANA201012
  • MANA201015
  • MANA201018
  • MANA201024
  • MANA201032
  • Grade:
  • MN 640 d
  • MN 640 d
  • MN 640 d
  • MN 640 d
  • MN 640 d
  • MN 640 d
  • MN 640 d
  • MN 640 d
  • MN 640 d
  • MN 640 dd
  • MN 640 dd
  • MN 640 dd
  • MN 640 dd
  • MN 640 dd
  • MN 640 dd
  • MN 640 dd
  • MN 640 dd
  • MN 640 dd
  • MN 640 de
  • MN 640 de
  • MN 640 de
  • MN 640 de
  • MN 640 de
  • MN 640 de
  • MN 640 de
  • MN 640 de
  • MN 640 de
  • MN 640 m
  • MN 640 m
  • MN 640 m
  • MN 640 m
  • MN 640 m
  • MN 640 m
  • MN 640 m
  • MN 640 m
  • MN 640 m
  • MN 640 md
  • MN 640 md
  • MN 640 md
  • MN 640 md
  • MN 640 md
  • MN 640 md
  • MN 640 md
  • MN 640 md
  • MN 640 md
  • MN 640 w
  • MN 640 w
  • MN 640 w
  • MN 640 w
  • MN 640 w
  • MN 640 w
  • MN 640 w
  • MN 640 w
  • MN 640 w
  • MN 640 we
  • MN 640 we
  • MN 640 we
  • MN 640 we
  • MN 640 we
  • MN 640 we
  • MN 640 we
  • MN 640 we
  • MN 640 we
  • Weight:
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 70 g/m²
  • 70 g/m²
  • 70 g/m²
  • 70 g/m²
  • 70 g/m²
  • 70 g/m²
  • 70 g/m²
  • 70 g/m²
  • 70 g/m²
  • 100 g/m²
  • 100 g/m²
  • 100 g/m²
  • 100 g/m²
  • 100 g/m²
  • 100 g/m²
  • 100 g/m²
  • 100 g/m²
  • 100 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • Thickness:
  • 0,17 mm
  • 0,17 mm
  • 0,17 mm
  • 0,17 mm
  • 0,17 mm
  • 0,17 mm
  • 0,17 mm
  • 0,17 mm
  • 0,17 mm
  • 0,16 mm
  • 0,16 mm
  • 0,16 mm
  • 0,16 mm
  • 0,16 mm
  • 0,16 mm
  • 0,16 mm
  • 0,16 mm
  • 0,16 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,22 mm
  • 0,22 mm
  • 0,22 mm
  • 0,22 mm
  • 0,22 mm
  • 0,22 mm
  • 0,22 mm
  • 0,22 mm
  • 0,22 mm
  • Particle retention:
  • 2 - 4 µm
  • 2 - 4 µm
  • 2 - 4 µm
  • 2 - 4 µm
  • 2 - 4 µm
  • 2 - 4 µm
  • 2 - 4 µm
  • 2 - 4 µm
  • 2 - 4 µm
  • -
  • -
  • -
  • -
  • -
  • -
  • -
  • -
  • -
  • 1 - 2 µm
  • 1 - 2 µm
  • 1 - 2 µm
  • 1 - 2 µm
  • 1 - 2 µm
  • 1 - 2 µm
  • 1 - 2 µm
  • 1 - 2 µm
  • 1 - 2 µm
  • 4 - 12 µm
  • 4 - 12 µm
  • 4 - 12 µm
  • 4 - 12 µm
  • 4 - 12 µm
  • 4 - 12 µm
  • 4 - 12 µm
  • 4 - 12 µm
  • 4 - 12 µm
  • 4 - 7 µm
  • 4 - 7 µm
  • 4 - 7 µm
  • 4 - 7 µm
  • 4 - 7 µm
  • 4 - 7 µm
  • 4 - 7 µm
  • 4 - 7 µm
  • 4 - 7 µm
  • 7 - 12 µm
  • 7 - 12 µm
  • 7 - 12 µm
  • 7 - 12 µm
  • 7 - 12 µm
  • 7 - 12 µm
  • 7 - 12 µm
  • 7 - 12 µm
  • 7 - 12 µm
  • 12 - 25 µm
  • 12 - 25 µm
  • 12 - 25 µm
  • 12 - 25 µm
  • 12 - 25 µm
  • 12 - 25 µm
  • 12 - 25 µm
  • 12 - 25 µm
  • 12 - 25 µm
  • Filtration speed:
  • 140 s (DIN 53137)
  • 140 s (DIN 53137)
  • 140 s (DIN 53137)
  • 140 s (DIN 53137)
  • 140 s (DIN 53137)
  • 140 s (DIN 53137)
  • 140 s (DIN 53137)
  • 140 s (DIN 53137)
  • 140 s (DIN 53137)
  • 100 s (DIN 53137)
  • 100 s (DIN 53137)
  • 100 s (DIN 53137)
  • 100 s (DIN 53137)
  • 100 s (DIN 53137)
  • 100 s (DIN 53137)
  • 100 s (DIN 53137)
  • 100 s (DIN 53137)
  • 100 s (DIN 53137)
  • 195 s (DIN 53137)
  • 195 s (DIN 53137)
  • 195 s (DIN 53137)
  • 195 s (DIN 53137)
  • 195 s (DIN 53137)
  • 195 s (DIN 53137)
  • 195 s (DIN 53137)
  • 195 s (DIN 53137)
  • 195 s (DIN 53137)
  • 27 s (DIN 53137)
  • 27 s (DIN 53137)
  • 27 s (DIN 53137)
  • 27 s (DIN 53137)
  • 27 s (DIN 53137)
  • 27 s (DIN 53137)
  • 27 s (DIN 53137)
  • 27 s (DIN 53137)
  • 27 s (DIN 53137)
  • 55 s (DIN 53137)
  • 55 s (DIN 53137)
  • 55 s (DIN 53137)
  • 55 s (DIN 53137)
  • 55 s (DIN 53137)
  • 55 s (DIN 53137)
  • 55 s (DIN 53137)
  • 55 s (DIN 53137)
  • 55 s (DIN 53137)
  • 9 s (DIN 53137)
  • 9 s (DIN 53137)
  • 9 s (DIN 53137)
  • 9 s (DIN 53137)
  • 9 s (DIN 53137)
  • 9 s (DIN 53137)
  • 9 s (DIN 53137)
  • 9 s (DIN 53137)
  • 9 s (DIN 53137)
  • 5 s (DIN 53137)
  • 5 s (DIN 53137)
  • 5 s (DIN 53137)
  • 5 s (DIN 53137)
  • 5 s (DIN 53137)
  • 5 s (DIN 53137)
  • 5 s (DIN 53137)
  • 5 s (DIN 53137)
  • 5 s (DIN 53137)
  • Ash content:
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
  • <0,01%
Spécifications
Product Family Options
Grade MN 640 d, circles
Informations sur le produit
  • Grade
    Size
    Disponibilité
    Prix
  • MN 640 d
    55 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 12,40 €
    12,40 €
    1 * 100 ST
       12,40 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    MANA205005
    Grade
    MN 640 d
    Weight
    85 g/m²
    Thickness
    0,17 mm
    Particle retention
    2 - 4 µm
    Filtration speed
    140 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 d
    70 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 16,40 €
    16,40 €
    1 * 100 ST
       16,40 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    512-0495
    Grade
    MN 640 d
    Weight
    85 g/m²
    Thickness
    0,17 mm
    Particle retention
    2 - 4 µm
    Filtration speed
    140 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 d
    90 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 24,00 €
    24,00 €
    1 * 100 ST
       24,00 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    512-0496
    Grade
    MN 640 d
    Weight
    85 g/m²
    Thickness
    0,17 mm
    Particle retention
    2 - 4 µm
    Filtration speed
    140 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 d
    110 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 33,70 €
    33,70 €
    1 * 100 ST
       33,70 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    512-0497
    Grade
    MN 640 d
    Weight
    85 g/m²
    Thickness
    0,17 mm
    Particle retention
    2 - 4 µm
    Filtration speed
    140 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 d
    125 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 39,20 €
    39,20 €
    1 * 100 ST
       39,20 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    515-9800
    Grade
    MN 640 d
    Weight
    85 g/m²
    Thickness
    0,17 mm
    Particle retention
    2 - 4 µm
    Filtration speed
    140 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 d
    150 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 54,40 €
    54,40 €
    1 * 100 ST
       54,40 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    MANA205015
    Grade
    MN 640 d
    Weight
    85 g/m²
    Thickness
    0,17 mm
    Particle retention
    2 - 4 µm
    Filtration speed
    140 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 d
    185 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 81,00 €
    81,00 €
    1 * 100 ST
       81,00 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    MANA205018
    Grade
    MN 640 d
    Weight
    85 g/m²
    Thickness
    0,17 mm
    Particle retention
    2 - 4 µm
    Filtration speed
    140 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 d
    240 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 137,00 €
    137,00 €
    1 * 100 ST
       137,00 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    MANA205024
    Grade
    MN 640 d
    Weight
    85 g/m²
    Thickness
    0,17 mm
    Particle retention
    2 - 4 µm
    Filtration speed
    140 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 d
    320 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 232,00 €
    232,00 €
    1 * 100 ST
       232,00 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    MANA205032
    Grade
    MN 640 d
    Weight
    85 g/m²
    Thickness
    0,17 mm
    Particle retention
    2 - 4 µm
    Filtration speed
    140 s (DIN 53137)
    Ash content
    <0,01%
Grade MN 640 dd, circles
Informations sur le produit
  • Grade
    Size
    Disponibilité
    Prix
  • MN 640 dd
    55 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 14,30 €
    14,30 €
    1 * 100 ST
       14,30 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    MANA206005
    Grade
    MN 640 dd
    Weight
    70 g/m²
    Thickness
    0,16 mm
    Particle retention
    -
    Filtration speed
    100 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 dd
    70 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 19,00 €
    19,00 €
    1 * 100 ST
       19,00 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    MANA206007
    Grade
    MN 640 dd
    Weight
    70 g/m²
    Thickness
    0,16 mm
    Particle retention
    -
    Filtration speed
    100 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 dd
    90 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 28,70 €
    28,70 €
    1 * 100 ST
       28,70 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    MANA206009
    Grade
    MN 640 dd
    Weight
    70 g/m²
    Thickness
    0,16 mm
    Particle retention
    -
    Filtration speed
    100 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 dd
    110 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 38,70 €
    38,70 €
    1 * 100 ST
       38,70 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    MANA206011
    Grade
    MN 640 dd
    Weight
    70 g/m²
    Thickness
    0,16 mm
    Particle retention
    -
    Filtration speed
    100 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 dd
    125 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 46,30 €
    46,30 €
    1 * 100 ST
       46,30 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    MANA206012
    Grade
    MN 640 dd
    Weight
    70 g/m²
    Thickness
    0,16 mm
    Particle retention
    -
    Filtration speed
    100 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 dd
    150 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 63,70 €
    63,70 €
    1 * 100 ST
       63,70 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    MANA206015
    Grade
    MN 640 dd
    Weight
    70 g/m²
    Thickness
    0,16 mm
    Particle retention
    -
    Filtration speed
    100 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 dd
    185 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 95,60 €
    95,60 €
    1 * 100 ST
       95,60 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    MANA206018
    Grade
    MN 640 dd
    Weight
    70 g/m²
    Thickness
    0,16 mm
    Particle retention
    -
    Filtration speed
    100 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 dd
    240 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 170,00 €
    170,00 €
    1 * 100 ST
       170,00 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    MANA206024
    Grade
    MN 640 dd
    Weight
    70 g/m²
    Thickness
    0,16 mm
    Particle retention
    -
    Filtration speed
    100 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 dd
    320 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 280,00 €
    280,00 €
    1 * 100 ST
       280,00 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    MANA206032
    Grade
    MN 640 dd
    Weight
    70 g/m²
    Thickness
    0,16 mm
    Particle retention
    -
    Filtration speed
    100 s (DIN 53137)
    Ash content
    <0,01%
Grade MN 640 de, circles
Informations sur le produit
  • Grade
    Size
    Disponibilité
    Prix
  • MN 640 de
    55 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 14,30 €
    14,30 €
    1 * 100 ST
       14,30 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    MANA207005
    Grade
    MN 640 de
    Weight
    100 g/m²
    Thickness
    0,20 mm
    Particle retention
    1 - 2 µm
    Filtration speed
    195 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 de
    70 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 19,00 €
    19,00 €
    1 * 100 ST
       19,00 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    MANA207007
    Grade
    MN 640 de
    Weight
    100 g/m²
    Thickness
    0,20 mm
    Particle retention
    1 - 2 µm
    Filtration speed
    195 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 de
    90 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 28,70 €
    28,70 €
    1 * 100 ST
       28,70 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    MANA207009
    Grade
    MN 640 de
    Weight
    100 g/m²
    Thickness
    0,20 mm
    Particle retention
    1 - 2 µm
    Filtration speed
    195 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 de
    110 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 38,70 €
    38,70 €
    1 * 100 ST
       38,70 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    MANA207011
    Grade
    MN 640 de
    Weight
    100 g/m²
    Thickness
    0,20 mm
    Particle retention
    1 - 2 µm
    Filtration speed
    195 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 de
    125 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 46,30 €
    46,30 €
    1 * 100 ST
       46,30 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    MANA207012
    Grade
    MN 640 de
    Weight
    100 g/m²
    Thickness
    0,20 mm
    Particle retention
    1 - 2 µm
    Filtration speed
    195 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 de
    150 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 63,70 €
    63,70 €
    1 * 100 ST
       63,70 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    MANA207015
    Grade
    MN 640 de
    Weight
    100 g/m²
    Thickness
    0,20 mm
    Particle retention
    1 - 2 µm
    Filtration speed
    195 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 de
    185 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 95,60 €
    95,60 €
    1 * 100 ST
       95,60 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    MANA207018
    Grade
    MN 640 de
    Weight
    100 g/m²
    Thickness
    0,20 mm
    Particle retention
    1 - 2 µm
    Filtration speed
    195 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 de
    240 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 170,00 €
    170,00 €
    1 * 100 ST
       170,00 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    MANA207024
    Grade
    MN 640 de
    Weight
    100 g/m²
    Thickness
    0,20 mm
    Particle retention
    1 - 2 µm
    Filtration speed
    195 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 de
    320 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 280,00 €
    280,00 €
    1 * 100 ST
       280,00 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    MANA207032
    Grade
    MN 640 de
    Weight
    100 g/m²
    Thickness
    0,20 mm
    Particle retention
    1 - 2 µm
    Filtration speed
    195 s (DIN 53137)
    Ash content
    <0,01%
Grade MN 640 m, circles
Informations sur le produit
  • Grade
    Size
    Disponibilité
    Prix
  • MN 640 m
    55 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 12,40 €
    12,40 €
    1 * 100 ST
       12,40 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    MANA203005
    Grade
    MN 640 m
    Weight
    85 g/m²
    Thickness
    0,20 mm
    Particle retention
    4 - 12 µm
    Filtration speed
    27 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 m
    70 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 16,40 €
    16,40 €
    1 * 100 ST
       16,40 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    512-0490
    Grade
    MN 640 m
    Weight
    85 g/m²
    Thickness
    0,20 mm
    Particle retention
    4 - 12 µm
    Filtration speed
    27 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 m
    90 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 24,00 €
    24,00 €
    1 * 100 ST
       24,00 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    MANA203009
    Grade
    MN 640 m
    Weight
    85 g/m²
    Thickness
    0,20 mm
    Particle retention
    4 - 12 µm
    Filtration speed
    27 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 m
    110 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 33,70 €
    33,70 €
    1 * 100 ST
       33,70 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    512-0491
    Grade
    MN 640 m
    Weight
    85 g/m²
    Thickness
    0,20 mm
    Particle retention
    4 - 12 µm
    Filtration speed
    27 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 m
    125 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 39,20 €
    39,20 €
    1 * 100 ST
       39,20 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    515-9710
    Grade
    MN 640 m
    Weight
    85 g/m²
    Thickness
    0,20 mm
    Particle retention
    4 - 12 µm
    Filtration speed
    27 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 m
    150 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 54,40 €
    54,40 €
    1 * 100 ST
       54,40 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    512-0492
    Grade
    MN 640 m
    Weight
    85 g/m²
    Thickness
    0,20 mm
    Particle retention
    4 - 12 µm
    Filtration speed
    27 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 m
    185 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 81,00 €
    81,00 €
    1 * 100 ST
       81,00 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    512-0493
    Grade
    MN 640 m
    Weight
    85 g/m²
    Thickness
    0,20 mm
    Particle retention
    4 - 12 µm
    Filtration speed
    27 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 m
    240 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 137,00 €
    137,00 €
    1 * 100 ST
       137,00 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    MANA203024
    Grade
    MN 640 m
    Weight
    85 g/m²
    Thickness
    0,20 mm
    Particle retention
    4 - 12 µm
    Filtration speed
    27 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 m
    320 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 232,00 €
    232,00 €
    1 * 100 ST
       232,00 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    MANA203032
    Grade
    MN 640 m
    Weight
    85 g/m²
    Thickness
    0,20 mm
    Particle retention
    4 - 12 µm
    Filtration speed
    27 s (DIN 53137)
    Ash content
    <0,01%
Grade MN 640 md, circles
Informations sur le produit
  • Grade
    Size
    Disponibilité
    Prix
  • MN 640 md
    55 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 12,40 €
    12,40 €
    1 * 100 ST
       12,40 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    MANA204005
    Grade
    MN 640 md
    Weight
    85 g/m²
    Thickness
    0,20 mm
    Particle retention
    4 - 7 µm
    Filtration speed
    55 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 md
    70 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 16,40 €
    16,40 €
    1 * 100 ST
       16,40 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    MANA204007
    Grade
    MN 640 md
    Weight
    85 g/m²
    Thickness
    0,20 mm
    Particle retention
    4 - 7 µm
    Filtration speed
    55 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 md
    90 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 24,00 €
    24,00 €
    1 * 100 ST
       24,00 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    MANA204009
    Grade
    MN 640 md
    Weight
    85 g/m²
    Thickness
    0,20 mm
    Particle retention
    4 - 7 µm
    Filtration speed
    55 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 md
    110 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 33,70 €
    33,70 €
    1 * 100 ST
       33,70 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    MANA204011
    Grade
    MN 640 md
    Weight
    85 g/m²
    Thickness
    0,20 mm
    Particle retention
    4 - 7 µm
    Filtration speed
    55 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 md
    125 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 39,20 €
    39,20 €
    1 * 100 ST
       39,20 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    512-0494
    Grade
    MN 640 md
    Weight
    85 g/m²
    Thickness
    0,20 mm
    Particle retention
    4 - 7 µm
    Filtration speed
    55 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 md
    150 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 54,40 €
    54,40 €
    1 * 100 ST
       54,40 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    MANA204015
    Grade
    MN 640 md
    Weight
    85 g/m²
    Thickness
    0,20 mm
    Particle retention
    4 - 7 µm
    Filtration speed
    55 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 md
    185 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 81,00 €
    81,00 €
    1 * 100 ST
       81,00 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    MANA204018
    Grade
    MN 640 md
    Weight
    85 g/m²
    Thickness
    0,20 mm
    Particle retention
    4 - 7 µm
    Filtration speed
    55 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 md
    240 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 137,00 €
    137,00 €
    1 * 100 ST
       137,00 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    MANA204024
    Grade
    MN 640 md
    Weight
    85 g/m²
    Thickness
    0,20 mm
    Particle retention
    4 - 7 µm
    Filtration speed
    55 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 md
    320 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 232,00 €
    232,00 €
    1 * 100 ST
       232,00 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    MANA204032
    Grade
    MN 640 md
    Weight
    85 g/m²
    Thickness
    0,20 mm
    Particle retention
    4 - 7 µm
    Filtration speed
    55 s (DIN 53137)
    Ash content
    <0,01%
Grade MN 640 w, circles
Informations sur le produit
  • Grade
    Size
    Disponibilité
    Prix
  • MN 640 w
    55 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 12,40 €
    12,40 €
    1 * 100 ST
       12,40 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    512-0084
    Grade
    MN 640 w
    Weight
    85 g/m²
    Thickness
    0,20 mm
    Particle retention
    7 - 12 µm
    Filtration speed
    9 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 w
    70 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 16,40 €
    16,40 €
    1 * 100 ST
       16,40 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    512-0486
    Grade
    MN 640 w
    Weight
    85 g/m²
    Thickness
    0,20 mm
    Particle retention
    7 - 12 µm
    Filtration speed
    9 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 w
    90 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 24,00 €
    24,00 €
    1 * 100 ST
       24,00 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    515-9700
    Grade
    MN 640 w
    Weight
    85 g/m²
    Thickness
    0,20 mm
    Particle retention
    7 - 12 µm
    Filtration speed
    9 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 w
    110 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 33,70 €
    33,70 €
    1 * 100 ST
       33,70 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    512-0487
    Grade
    MN 640 w
    Weight
    85 g/m²
    Thickness
    0,20 mm
    Particle retention
    7 - 12 µm
    Filtration speed
    9 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 w
    125 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 39,20 €
    39,20 €
    1 * 100 ST
       39,20 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    515-9701
    Grade
    MN 640 w
    Weight
    85 g/m²
    Thickness
    0,20 mm
    Particle retention
    7 - 12 µm
    Filtration speed
    9 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 w
    150 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 54,40 €
    54,40 €
    1 * 100 ST
       54,40 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    512-0488
    Grade
    MN 640 w
    Weight
    85 g/m²
    Thickness
    0,20 mm
    Particle retention
    7 - 12 µm
    Filtration speed
    9 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 w
    185 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 81,00 €
    81,00 €
    1 * 100 ST
       81,00 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    MANA202018
    Grade
    MN 640 w
    Weight
    85 g/m²
    Thickness
    0,20 mm
    Particle retention
    7 - 12 µm
    Filtration speed
    9 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 w
    240 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 137,00 €
    137,00 €
    1 * 100 ST
       137,00 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    512-0489
    Grade
    MN 640 w
    Weight
    85 g/m²
    Thickness
    0,20 mm
    Particle retention
    7 - 12 µm
    Filtration speed
    9 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 w
    320 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 232,00 €
    232,00 €
    1 * 100 ST
       232,00 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    MANA202032
    Grade
    MN 640 w
    Weight
    85 g/m²
    Thickness
    0,20 mm
    Particle retention
    7 - 12 µm
    Filtration speed
    9 s (DIN 53137)
    Ash content
    <0,01%
Grade MN 640 we, circles
Informations sur le produit
  • Grade
    Size
    Disponibilité
    Prix
  • MN 640 we
    55 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 12,40 €
    12,40 €
    1 * 100 ST
       12,40 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    MANA201005
    Grade
    MN 640 we
    Weight
    85 g/m²
    Thickness
    0,22 mm
    Particle retention
    12 - 25 µm
    Filtration speed
    5 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 we
    70 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 16,40 €
    16,40 €
    1 * 100 ST
       16,40 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    MANA201007
    Grade
    MN 640 we
    Weight
    85 g/m²
    Thickness
    0,22 mm
    Particle retention
    12 - 25 µm
    Filtration speed
    5 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 we
    90 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 24,00 €
    24,00 €
    1 * 100 ST
       24,00 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    MANA201009
    Grade
    MN 640 we
    Weight
    85 g/m²
    Thickness
    0,22 mm
    Particle retention
    12 - 25 µm
    Filtration speed
    5 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 we
    110 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 33,70 €
    33,70 €
    1 * 100 ST
       33,70 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    MANA201011
    Grade
    MN 640 we
    Weight
    85 g/m²
    Thickness
    0,22 mm
    Particle retention
    12 - 25 µm
    Filtration speed
    5 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 we
    125 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 39,20 €
    39,20 €
    1 * 100 ST
       39,20 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    MANA201012
    Grade
    MN 640 we
    Weight
    85 g/m²
    Thickness
    0,22 mm
    Particle retention
    12 - 25 µm
    Filtration speed
    5 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 we
    150 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 54,40 €
    54,40 €
    1 * 100 ST
       54,40 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    MANA201015
    Grade
    MN 640 we
    Weight
    85 g/m²
    Thickness
    0,22 mm
    Particle retention
    12 - 25 µm
    Filtration speed
    5 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 we
    185 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 81,00 €
    81,00 €
    1 * 100 ST
       81,00 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    MANA201018
    Grade
    MN 640 we
    Weight
    85 g/m²
    Thickness
    0,22 mm
    Particle retention
    12 - 25 µm
    Filtration speed
    5 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 we
    240 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 137,00 €
    137,00 €
    1 * 100 ST
       137,00 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    MANA201024
    Grade
    MN 640 we
    Weight
    85 g/m²
    Thickness
    0,22 mm
    Particle retention
    12 - 25 µm
    Filtration speed
    5 s (DIN 53137)
    Ash content
    <0,01%
  • MN 640 we
    320 mm
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 232,00 €
    232,00 €
    1 * 100 ST
       232,00 €
    Spécifications :
    Plus
    Specifications
    Cat. No.
    MANA201032
    Grade
    MN 640 we
    Weight
    85 g/m²
    Thickness
    0,22 mm
    Particle retention
    12 - 25 µm
    Filtration speed
    5 s (DIN 53137)
    Ash content
    <0,01%
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.