Orders LinkContactUsLinkComponent
Cimarec™ VARIOMAG® i Telemodul Controller
Cimarec™ VARIOMAG® i Telemodul Controller
Telemodul stirrer controller, 230 V, operating at 0...40 °C (80% relative humidity), 96×63×50 mm
Catalog # HPLT50119118
Cimarec™ VARIOMAG® i Telemodul Controller
Catalog # HPLT50119118
Cimarec™ VARIOMAG® i Telemodul Controller
Cimarec™ VARIOMAG® i Telemodul Controller
Telemodul stirrer controller, 230 V, operating at 0...40 °C (80% relative humidity), 96×63×50 mm
Price
382,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Save space with Thermo Scientific™ Cimarec™ i Telemodul controllers, including Micro, Mini, Compact and Maxi stirrers.
  • Provide stirring power for small to medium volumes of low-viscosity fluids
  • Recommended for single point stirring up to 250 ml
Ordering information: Order as replacement controller only.
Specifications
  • Description:
    Telemodul stirrer controller, 230 V, operating at 0...40 °C (80% relative humidity), 96×63×50 mm
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.