VWR® Mega Racks® Tube Racks for 28 to 32 mm Tubes
Fournisseur: VWR Collection
New Product
Certificats
À propos de cet article
High-capacity storage for 50 ml tubes.
- Sturdy and lightweight for easy and secure transport of tubes
- Place different test groupings into one rack and eliminate need for multiple tube racks
- Alphanumeric identification for quick location and retrieval of tubes
- Autoclavable
These sturdy and lightweight mega tube racks are great for high throughput laboratories requiring large sample storage. They allow you to place different test groupings into one rack, eliminating the need for multiple tube racks. These racks also feature alphanumeric identification for quick location and retrieval of tubes. Available in various colors and sizes.
Frequently Bought Together