p38 MAP Kinase Inhibitor VIII ≥97% (by HPLC), Calbiochem®, Sigma-Aldrich®
Catalog # 506163-5MG
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pack type:Plastic ampoule
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:415.71 g/mol
:M-Clarity™ quality level = MQ100
Storage Temperature:Refrigerator
CAS Number:321351-00-2
Formula:C₂₀H₁₆BrClN₂O