Order Entry
Belgium
ContactUsLinkComponent
SDS-PAGE Sample Buffer, 4X, (2-Mercaptoethanol-free) for Electrophoresis
SDS-PAGE Sample Buffer, 4X, (2-Mercaptoethanol-free) for Electrophoresis
Catalog # TCIAB6104-20ML
Supplier:  TCI
CAS Number:  
SDS-PAGE Sample Buffer, 4X, (2-Mercaptoethanol-free) for Electrophoresis
Catalog # TCIAB6104-20ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    20 mL
  • Storage temperature:
    Room Temperature (Recommended in a cool and dark place, <15°C)
  • Appearance:
    Purple to dark purple clear liquid to cloudy liquid.

Specifications

About this item

This product contains 8% SDS, 40% glycerol, 0.02% bromophenol blue and 0.25 M Tris HCl, pH approx. 6.8. Does not contain 2-Mercaptoethanol.

SDS-PAGE Sample Buffer is specially formulated for protein sample preparation based on the Laemmli Method of SDS-PAGE system.

Preparation of non-reducing sample
A protein sample is mixed with the 4X SDS-PAGE sample buffer (3:1). If necessary, heating the mix solution and then cool down the tube at room temperature. The solution is ready for SDS-PAGE.

Preparation of reducing sample (reducing with 2-Mercaptoethanol)
Add 20 μl of 2-Mercaptoethanol per 80 μl of 4X SDS-PAGE sample buffer. Mix thoroughly. Then a protein sample is mixed with the sample buffer (3:1) and heating to 95 - 100 °C for 5 min. Cool down the tube at room temperature. The solution is ready for SDS-PAGE.
Reducing reagents: 2-Mercaptoethanol [for Electrophoresis], DL-Dithiothreitol [for Electrophoresis], etc.

Caution: Please use after bringing back to normal temperature.