NeuroFluor™ NeuO Membrane-Permeable Fluorescent Probe
Catalog # STMC01801
Supplier: STEMCELL Technologies
CAS Number:
Specifications
- Pk:0,1 mL
- Application:
- Description:NeuroFluor™ NeuO
- Environmentally Preferable:
Specifications
About this item
Membrane-permeable fluorescent probe for the detection of live neurons - 0,1 ml.
- Enables selective labelling of mouse, rat, or human neurons without fixation
- Can be used to confirm neuronal differentiation of human pluripotent stem cell-derived neural progenitor cells
- Can be used to label neurons in the presence of other brain cells
- Non-toxic and non-permanent
- Simple and rapid labelling protocol
NeuroFluor™ NeuO is a membrane-permeable fluorescent probe that selectively labels primary and pluripotent stem cell-derived neurons in the presence of other brain cells (J C Er et al. Angew Chem Int Ed, 2015). Cells labeled with NeuroFluor™ NeuO can be visualised using fluorescent imaging. labelling with this probe is non-permanent; it can be washed off, providing unlabeled, viable cells for downstream applications. For additional information, see References.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').