ALDEFLUOR™ Kit
ALDEFLUOR™ kit for the identification, evaluation and isolation of stem and progenitor cells expressing high levels of ALDH - 1 kit.
- Identify viable cells with elevated ALDH activity
- Use to identify and isolate normal and malignant stem and progenitor cells across a wide variety of tissues
Identify and isolate viable cells expressing aldehyde dehydrogenase (ALDH) with the ALDEFLUOR™ kit. Compared to traditional methods, this assay does not require antibody staining.
High ALDH expression has been reported for normal and cancer precursor cells of various lineages. The ALDEFLUOR™ assay is a widely published non-immunological method for the detection of ALDH-bright (ALDHbr) cells and can be used to detect cancer cells in hematopoietic, mammary, endothelial, mesenchymal, neural, and other tissues.
ALDEFLUOR™ DEAB Reagent and ALDEFLUOR™ assay buffer, included with the kit, support the optimal performance of the assay and are also available for purchase individually.
The kit is compatible with standard flow cytometers for performing downstream analysis of ALDHbr cells and with standard cell sorters for further purification and characterisation.
CAS No: 7647-01-0
: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
- Pk:1 KIT
- Description:ALDEFLUOR™ kit