ContactUsLinkComponent
Cell Navigator™ Cell Plasma Membrane Staining Kit, Green
Cell Navigator™ Cell Plasma Membrane Staining Kit, Green
Catalog # STMC100-0835
CAS Number:  
undefined
Cell Navigator™ Cell Plasma Membrane Staining Kit, Green
Catalog # STMC100-0835
CAS Number:  
Specifications
  • Pk:
    500 Tests
  • Description:
    Cell Navigator™ Cell plasma membrane staining kit, green
Specifications
About this item
For staining cell plasma membranes in fixed cells - 500 tests.
Cell Navigator™ Cell Plasma Membrane Staining Kits contain a fluorescently labeled dye for staining the plasma membrane in fixed cells. The fluorescently labeled dye stains the plasma membrane without showing cell-type differences as observed with lectins. The dye can be used as a segmentation tool during high-content screening. Cell Navigator™ Cell Plasma Membrane Staining Kit, Deep Red includes Cellpaint™ Deep Red, Live Cell Staining Buffer, and DMSO. Cell Navigator™ Cell Plasma Membrane Staining Kit, Green includes Cellpaint™ Green, Live Cell Staining Buffer, and DMSO.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').