ContactUsLinkComponent
ArciTect™ sgRNA
ArciTect™ sgRNA
Catalog # STMC200-0013
CAS Number:  
undefined
ArciTect™ sgRNA
Catalog # STMC200-0013
CAS Number:  
Specifications
  • Pk:
    4 Nmo
  • Description:
    ArciTect™ sgRNA
Specifications
About this item
Custom-designed single guide RNA for CRISPR-Cas9 genome editing.
ArciTect™ sgRNA is a custom single guide RNA (sgRNA) for CRISPR-Cas9 genome editing. ArciTect™ sgRNA contains a user-specified 19- to 21-base sequence complementary to the target genomic location. ArciTect™ sgRNA must be designed directly upstream of a protospacer adjacent motif (PAM) site (5’-NGG-3’). It contains 2'-O-methyl and phosphorothioate modifications at the first two 5' and 3' terminal residues for optimal stability and editing efficiency.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').