ContactUsLinkComponent
ArciTect™ Annealing Buffer (5X)
ArciTect™ Annealing Buffer (5X)
Catalog # STMC76020
CAS Number:  
undefined
ArciTect™ Annealing Buffer (5X)
Catalog # STMC76020
CAS Number:  
Specifications
  • Pk:
    1 mL
  • Cell specific:
    Pluripotent stem cells
  • Description:
    ArciTect™ Annealing buffer (5X)
Specifications
About this item
For guide RNA generation in CRISPR-Cas9 genome editing.
ArciTect™ Annealing Buffer (5X) is required to anneal ArciTect™ tracrRNA and ArciTect™ crRNA to form a guide RNA (gRNA) duplex. This product is a refill component for ArciTect™ tracrRNA kit.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').