Winkler Electrodes
Supplier: CHEMPUR
New Product
Certificates
About this item
Electrode for electrogravimetrical identification of Cu and Zn by Winkler method.
- Made of platinum/iridium 90/10
This electrode consists of a slotted net cylinder in which a with spiral is concentrically arranged.
Frequently Bought Together