Orders LinkContactUsLinkComponent
Label Rolls, Core2Write
Supplier:   VELTEK ASSOCIATES
Label Rolls, Core2Write
Supplier:  VELTEK ASSOCIATES
Certificates
Label Rolls, Core2Write
Label Rolls, Core2Write
10 Options Available Below
Product Details & Documents
Constructed using CleanPrint 10, Core2Write labels are extremely durable, smear and smudge proof, chemical resistant, and remain pliable in extreme temperatures. All labels can be customised or left blank.
  • Always retain critical documentation, pre-printed and hand-written
  • Extremely durable
  • Abrasion and tear resistant
  • Smear resistant
  • Can withstand multiple chemicals
  • Can withstand a wide temperature range (–70 to 200 ℃)
  • Fully customisable by size, color, pre-printing
All roll labels come with a three inch plastic core. Can be applied to all types of surfaces: glass, poly, stainless steel. Can be autoclaved.
 : Also available in sheets (sold separately).
Product Family Options
Non-sterile
Product Information
  • Colour
    Dimensions
    Packed
    Availability
    Price
  • VELTC2WR2X1BLK01
    Core2Write label, non-sterile
    Core2Write label, non-sterile
    White
    50,8×25,4 mm
    1000 labels/roll, 4 rolls/case
    Log in for availability
    Log in for availability
    1 * 4.000 ST
    1 * 4.000 ST - 3 930,00 €
    3 930,00 €
    1 * 4.000 ST
       3 930,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VELTC2WR2X1BLK01
  • VELTC2WR3X1BLK01
    Core2Write label, non-sterile
    Core2Write label, non-sterile
    White
    76,2×25,4 mm
    1000 labels/roll, 4 rolls/case
    Log in for availability
    Log in for availability
    1 * 4.000 ST
    1 * 4.000 ST - 3 930,00 €
    3 930,00 €
    1 * 4.000 ST
       3 930,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VELTC2WR3X1BLK01
  • VELTC2WR3X1.5BLK01
    Core2Write label, non-sterile
    Core2Write label, non-sterile
    White
    76,2×38,1 mm
    1000 labels/roll, 4 rolls/case
    Log in for availability
    Log in for availability
    1 * 4.000 ST
    1 * 4.000 ST - 3 930,00 €
    3 930,00 €
    1 * 4.000 ST
       3 930,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VELTC2WR3X1.5BLK01
  • VELTC2WR4X2BLK01
    Core2Write label, non-sterile, irradiated
    Core2Write label, non-sterile, irradiated
    White
    101,6×50,8 mm
    1000 labels/roll, 4 rolls/case
    Log in for availability
    Log in for availability
    1 * 4.000 ST
    1 * 4.000 ST - 3 930,00 €
    3 930,00 €
    1 * 4.000 ST
       3 930,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VELTC2WR4X2BLK01
  • VELTC2WR4X3BLK01
    Core2Write label, non-sterile
    Core2Write label, non-sterile
    White
    101,6×76,2 mm
    1000 labels/roll, 4 rolls/case
    Log in for availability
    Log in for availability
    1 * 4.000 ST
    1 * 4.000 ST
    Specifications:
    More
    Specifications
    Cat. No.
    VELTC2WR4X3BLK01
Sterile
Product Information
  • Colour
    Dimensions
    Packed
    Availability
    Price
  • VELTC2WR2X1BLK02
    Core2Write label, sterile, irradiated
    Core2Write label, sterile, irradiated
    White
    50,8×25,4 mm
    1000 labels/roll, 4 rolls/case
    Log in for availability
    Log in for availability
    1 * 4.000 ST
    1 * 4.000 ST - 4 510,00 €
    4 510,00 €
    1 * 4.000 ST
       4 510,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VELTC2WR2X1BLK02
  • VELTC2WR3X1.5BLK02
    Core2Write label, sterile, irradiated
    Core2Write label, sterile, irradiated
    White
    76,2×38,1 mm
    1000 labels/roll, 4 rolls/case
    Log in for availability
    Log in for availability
    1 * 4.000 ST
    1 * 4.000 ST - 4 510,00 €
    4 510,00 €
    1 * 4.000 ST
       4 510,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VELTC2WR3X1.5BLK02
  • VELTC2WR3X1BLK02
    Core2Write label, sterile, irradiated
    Core2Write label, sterile, irradiated
    White
    76,2×25,4 mm
    1000 labels/roll, 4 rolls/case
    Log in for availability
    Log in for availability
    1 * 4.000 ST
    1 * 4.000 ST - 4 510,00 €
    4 510,00 €
    1 * 4.000 ST
       4 510,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VELTC2WR3X1BLK02
  • VELTC2WR4X2BLK02
    Core2Write label, sterile, irradiated
    Core2Write label, sterile, irradiated
    White
    101,6×50,8 mm
    1000 labels/roll, 4 rolls/case
    Log in for availability
    Log in for availability
    1 * 4.000 ST
    1 * 4.000 ST - 4 510,00 €
    4 510,00 €
    1 * 4.000 ST
       4 510,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VELTC2WR4X2BLK02
  • VELTC2WR4X3BLK02
    Core2Write label, sterile, irradiated
    Core2Write label, sterile, irradiated
    White
    101,6×76,2 mm
    1000 labels/roll, 4 rolls/case
    Log in for availability
    Log in for availability
    1 * 4.000 ST
    1 * 4.000 ST - 4 510,00 €
    4 510,00 €
    1 * 4.000 ST
       4 510,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VELTC2WR4X3BLK02