Presentation Folder (for Cutting Tools)
Presentation folder
N° de catalogue MARO900852.08
Fournisseur: MARTOR ARGENTAX
Jump to:
Product Details & Documents
This folder is convient for large selection of cutting tools.
- Dimension (W×D×H) : 40×250×330 mm
- Basic material: Nylon
- Weight product: 2,420 g
This folder is designed for transport cutting tools safely and present a convincing demonstration. Inside presentation folder contains high-quality safety knives and scissors at hand.
Specifications
- Description:Presentation folder
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



