Haloperidol Decanoate, Reference Standard
Catalog # EDQMY0002434
Supplier: EDQM
New Product
CAS Number:
Specifications
- Pack type:Vial
- Pk:15 mg
- Standards:European Pharmacopoeia Reference Standard
- Description:Haloperidol decanoate for system suitability CRS
- Storage conditions:5 °C (±3 °C)
- GHS signal:Warning
Specifications
About this item
Haloperidol decanoate for system suitability CRS is a reference standard for laboratory tests as prescribed in the European Pharmacopoeia only.