Order Entry
Belgium
Orders LinkContactUsLinkComponent
Drip Tray

Drip Tray

Security drip tray for MiniMix®, Stainless steel, 0,22 kg, 175×100×20 mm mm
Catalog # INSC016000
Supplier:   INTERSCIENCE
Drip Tray
Catalog # INSC016000
Supplier:  INTERSCIENCE
Drip Tray

Security drip tray for MiniMix®, Stainless steel, 0,22 kg, 175×100×20 mm mm

Price
149,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Capacity:
    0,22 kg
  • Description:
    Security drip tray for MiniMix®
  • Material:
    Stainless steel
  • W×D×H:
    175×100×20 mm mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.