Order Entry
Belgium
ContactUsLinkComponent
Ubiquinol
Ubiquinol
New Product
Certificates
Ubiquinol
New Product

Specifications

  • CAS No.:
    992-78-9
  • Shipping conditions:
    Dry Ice
  • Formula:
    C₅₉ H₉₂ O₄
  • Storage temperature:
    −86 °C

Specifications

Frequently Bought Together