Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Media for Microbiology
Prepared Media for Microbiology
Sabouraud Dextrose Agar (Irradiated)
Sabouraud Dextrose Agar (Irradiated)
Sabouraud Dextrose Agar (Irradiated)
Catalog # LIOF10114S
Supplier:
LIOFILCHEM
New Product
CAS Number:
Expand image
Sabouraud Dextrose Agar (Irradiated)
Catalog # LIOF10114S
Supplier:
LIOFILCHEM
New Product
CAS Number:
Specifications
Pk:
20
Description:
Sabouraud dextrose agar, 90 mm plate
Size:
30 ml
Specifications
About this item
Product Details
Product Details