Specpure® Copper, Oil Based Standard Solution, Cu 1000 µg/g
Catalog # 47276.18
Fournisseur: Thermo Fisher Scientific
Numéro CAS:
Spécifications
- Concentration:1000 µg/g
- Matrix:oil
- Pk:50 g
- MDL No.:MFCD00010965
- Element:Copper
Spécifications
À propos de cet article
Copper, Oil based standard solution, Specpure|r, Cu 1000^mg/g
Physical form: Liquid
MDL No.: MFCD00010965