Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Specpure® Interference Check Standard AB(ICS-AB) Solution

Interference check standard AB(ICS-AB) solution, 5% HNO₃/tr. (CHOH)₂(COOH)₂, 100 mL
Catalog # 41730.AE
Specpure® Interference Check Standard AB(ICS-AB) Solution
Catalog # 41730.AE
CAS Number:  
Specpure® Interference Check Standard AB(ICS-AB) Solution
Specpure® Interference Check Standard AB(ICS-AB) Solution

Interference check standard AB(ICS-AB) solution, 5% HNO₃/tr. (CHOH)₂(COOH)₂, 100 mL

CAS Number:  
Price
333,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

UN No.: UN3264
MDL No.: MFCD00147272

Specifications

Specifications

  • Matrix:
    5% HNO₃/tr. (CHOH)₂(COOH)₂
  • Pk:
    100 mL
  • Description:
    Interference check standard AB(ICS-AB) solution
  • UN No.:
    3264
  • Cat. No.:
    41730.AE
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00147272

Specifications