Order Entry
Belgium
ContactUsLinkComponent
Wiva™ VAT-5 Infinity Disposal Container - Polypropylene
Wiva™ VAT-5 Infinity Disposal Container - Polypropylene
Catalog # SEMA12178
Supplier:  SEMADENI
New Product
Wiva™ VAT-5 Infinity Disposal Container - Polypropylene
Catalog # SEMA12178
Supplier:  SEMADENI
New Product

Specifications

  • Colour:
    Grey
  • Description:
    Safety waste jug VAT-5, 30 L, dimension: 400×333,5×31 mm
  • Material:
    PP
  • Environmentally Preferable:
  • Pk:
    1

Specifications

About this item

Disposal container for clinical waste, chemical waste, and hazardous waste in laboratories.

  • Made from 100% recycled plastic
  • This reduces CO₂ emissions during production by 52%

The intelligent lid design offers a large number of advantages: Pressing gently on the lid causes the container to close, forming an odor-proof, airtight seal (day lid function); pressing firmly causes the container to hermetically seal for transport and disposal. The lid can be hung on the side of the container during use.

The click lid (item SEMA8687) has a round inner lid. Especially in places where the container is left standing for long periods of time and smaller amounts of waste are discarded, the round disposal opening can be conveniently opened and closed again. This significantly reduces the leakage of odor emissions. To lock the inner lid securely, this should be turned to the stop and then pressed down.

The containers are resistant to chemicals, heat and cold, and are very stable and drop-proof. They have two recessed and rounded handles.

Disposal containers are not suitable for transporting liquids; when they are securely closed for transport, the covers can no longer be opened. The containers are made from environmentally-friendly materials. Please observe the applicable local regulations for the disposal of hazardous waste. Cost effective purchase and transport costs; light and easy to handle; reduced space requirement because of square shape, stackable and palettable on top of each other.

Ordering information: Without lid. Suitable lids must be ordered separately.