Supelco® PESTANAL® Organic Reference Standard, Clothianidin-d₃
Catalog # 56816-50MG
Supplier: Merck
CAS Number:
Specifications
- Pk:50 mg
- Standards:PESTANAL®, analytical standard
- CAS No.:1262776-24-8
- Description:Clothianidin-d₃
- Storage conditions:2...30 °C
- Hazard class description:H302 - H410
- Shipping conditions:STD
- MDL No.:MFCD17019117
- M-Clarity quality level:MQ100
- Synonym(s):(E)-1-(2-Chloro-5-thiazolylmethyl)-3-(methyl-d₃)-2-nitroguanidine
- Molecular weight:252,70
Specifications
About this item
- GHS signal: GHS07, GHS09