Supelco® PESTANAL® Organic Reference Standard, Prochloraz
Catalog # 45631-250MG
Supplier: Merck
CAS Number:
Specifications
- Pk:250 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:67747-09-5
- Description:Prochloraz
- Storage conditions:2...30 °C
- Hazard class description:H302 - H410
- Shipping conditions:STD
- MDL No.:MFCD00078735
- M-Clarity quality level:MQ100
- Formula:C₁₅H₁₆Cl₃N₃O₂
- Molecular weight:376,67
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS07, GHS09
Caution: P264 - P270 - P273 - P301 + P312 - P391 - P501