Supelco® PESTANAL® Organic Reference Standard, Atrazine-Desethyl-Desisopropyl-2-Hydroxy
45613-250MG
: Merck
:
- Pk:250 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:645-92-1
- Description:Atrazine-desethyl-desisopropyl-2-hydroxy
- Storage conditions:2...30 °C
- Shipping conditions:STD
- MDL No.:MFCD00023186
- M-Clarity quality level:MQ100
- Synonym(s):4,6-Diamino-2-hydroxy-1,3,5-triazine; Ammeline
- Formula:C₃H₅N₅O
- Molecular weight:127,10
- Flash point:Not applicable